Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Naphthalene
Supelco® Organic Reference Standard, Naphthalene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Naphthalene
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    91-20-3
  • Storage conditions:
    RT
  • Hazard class description:
    H228 - H302 - H351 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00001742
  • M-Clarity quality level:
    MQ100
  • Melting point:
    79.5 - 81.0 °C; 80 - 82 °C (lit.)
  • Molecular weight:
    128,17
  • Flash point:
    78,5 °C - Closed cup
  • Boiling point:
    218 °C (lit.)

Specifications

Related Information

Frequently Bought Together