2,6-Dimethoxynaphthalene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5486-55-5
:
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00004063
:188.23 g/mol
Melting Pt:152…154 °C
Storage Temperature:Ambient
EINECS:226-816-9
Frequently Bought Together