
Octafluoronaphthalene 96%
Catalog # ACRO291530010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:85 to 90 °C
- GC:≥95 %
Specification Test Results
About this item
:
:272.1 g/mol
Boiling Pt:238 °C (1013 hPa)
Melting Pt:85…90 °C
Density:2,069 g/cm³ (-70 °C)
Storage Temperature:Ambient
EINECS:206-239-9
:GHS07
CAS Number:313-72-4
Formula:C₁₀F₈
MDL Number:MFCD00014307