2-Naphthaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 66-99-9 Molecular Formula: C11H8O Formula Weight: 156.18 Melting Point: 59-62° Boiling Point: 160°/19mm MDL No.: MFCD00004094
Synonyms: β-Naphthaldehyde
- CAS Number:66-99-9
- Formula:C₁₁H₈O
- MDL Number:MFCD00004094
- Molecular Weight:156.18 g/mol
- Boiling Pt:160 °C (19 mmHg)
- Melting Pt:59…62 °C
- Density:1.0775 g/cm³ (99 °C)
- Storage Temperature:Ambient
- EINECS:200-640-2
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 66-99-9 Molecular Formula: C11H8O Formula Weight: 156.18 Melting Point: 59-62° Boiling Point: 160°/19mm MDL No.: MFCD00004094
Synonyms: β-Naphthaldehyde
- CAS Number:66-99-9
- Formula:C₁₁H₈O
- MDL Number:MFCD00004094
- Molecular Weight:156.18 g/mol
- Boiling Pt:160 °C (19 mmHg)
- Melting Pt:59…62 °C
- Density:1.0775 g/cm³ (99 °C)
- Storage Temperature:Ambient
- EINECS:200-640-2
Documents
Organic Building Blocks and Intermediates



