Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Naphthaldehyde 98%

2-Naphthaldehyde 98%

Irritant
2-Naphthaldehyde 98%
Certificates
2-Naphthaldehyde 98%

3 Options Available Below

Product Details & Documents
Synonyms:  β-Naphthaldehyde
  • CAS Number:66-99-9
  • Formula:C₁₁H₈O
  • MDL Number:MFCD00004094
  • Molecular Weight:156.18 g/mol
  • Boiling Pt:160 °C (19 mmHg)
  • Melting Pt:58…62 °C
  • Density:1,0775 g/cm³ (99 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-640-2
  • UN:3335
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Off-white to yellow to light beige Powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 62 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 34,40 €
    34,40 €
    1 * 5 g
       34,40 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 112,00 €
    112,00 €
    1 * 25 g
       112,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 411,00 €
    411,00 €
    1 * 100 g
       411,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.