Order Entry
Italy
ContactUsLinkComponent
1-Bromonaphthalene for synthesis, Sigma-Aldrich®
Irritant
1-Bromonaphthalene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Irritant
1-Bromonaphthalene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    207.07 g/mol
  • Boiling Point:
    281 °C (1013 hPa)
  • Vapor Pressure:
    0.013 hPa (20 °C)
  • Flash Point:
    67 °C
  • Refractive Index:
    1.6576 (20 °C, 589 nm)
  • Density:
    1.48 g/cm3 (20 °C)
  • Synonyms:
    α-Naphthyl bromide
  • Hill Formula:
    C10H7Br

Specification Test Results

Related Information

Frequently Bought Together