

Osmium tetraoxide for microscopy, Sigma-Aldrich®
Catalog # 1.24505.0000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Solubility:65 g/l (20 °C)
- Melting Point:40 °C
- Molar Mass:254.19 g/mol
- Bulk Density:1500 kg/m3
- Boiling Point:130 °C (1013 hPa)
- Vapor Pressure:10 hPa (20 °C)
- Density:4.91 g/cm3 (20 °C)
- pH:6 (25 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:254.23 g/mol
Merck Index:13,06962
Boiling Pt:130 °C (1013 hPa)
Melting Pt:40 °C
Density:1.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:244-058-7
UN:2471
ADR:6.1,
:GHS06
:GHS05
CAS Number:20816-12-0
Formula:O₄Os
MDL Number:MFCD00011150