




Chromium(VI) oxide analytical reagent, Supelco®
Catalog # 1.00229.0250
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:1854 g/l (20 °C)
- Melting Point:197 °C
- Molar Mass:99.99 g/mol
- Bulk Density:900 kg/m3
- Density:2.7 g/cm3 (20 °C)
- pH:- 1 (100 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:99.99 g/mol
Merck Index:13,02256
Melting Pt:195 °C
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-607-8
UN:1463
ADR:5.1 (6.1, 8),II
:M-Clarity™ quality level = MQ300
REACH:01-2119458868-17
:GHS03
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:1333-82-0
Formula:CrO₃
MDL Number:MFCD00010952