
Chromium(III) oxide ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1308-38-9
:
Formula:Cr₂O₃
MDL Number:MFCD00010949
:151.99 g/mol
Merck Index:14,2234
Boiling Pt:3000 °C
Melting Pt:2435 °C
Density:5.22
EINECS:215-160-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together