Silver nitrate 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
Boiling Pt:444
Melting Pt:210
Density:4.35 g/cm³ (20 °C)
Molecular Weight:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
ADR:5.1 II
Merck Index:12,08661
UN:1493
REACH:01-2119513705-43
:GHS03
:GHS07
:GHS05
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.8 - 100.5 %
- Clarity of solution:Passes test ACS
- Free acid:Passes test ACS
- Identification (Ag):Passes test Ph.Eur.
- Identification (NO3):Passes test Ph.Eur.
- Solution in water:Passes test
- Appearance of solution:Passes test Ph.Eur.
- Acidity or alkalinity:Passes test Ph.Eur.
- Aluminium, lead, copper and bismuth:Passes test Ph.Eur.
- Foreign salts:≤ 0.3 %
- Insolubility in water:≤ 50 ppm
- Not precipitated by HCl (as SO4):≤ 100 ppm
- Cl (Chloride):≤ 5 ppm
- SO4 (Sulphate):≤ 20 ppm
- Ca (Calcium):≤ 10 ppm
- Cd (Cadmium):≤ 1 ppm
- Cu (Copper):≤ 2 ppm
- Fe (Iron):≤ 2 ppm
- K (Potassium):≤ 50 ppm
- Mg (Magnesium):≤ 5 ppm
- Mn (Manganese):≤ 5 ppm
- Ni (Nickel):≤ 5 ppm
- Pb (Lead):≤ 10 ppm
- Zn (Zinc):≤ 1 ppm
- Conforms to Reag. Ph.Eur./ACS:Passes test
Specification Test Results
Related Information
Frequently Bought Together