Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
Selenium dioxide for synthesis, sublimated, Sigma-Aldrich®
 :  Merck
 
 
 
 
Selenium dioxide for synthesis, sublimated, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    384 g/l (14°C)
  • Molar Mass:
    1500 kg/m3
  • Bulk Density:
    1500 kg/m3
  • Vapor Pressure:
    16 hPa (70 °C)
  • pH:
    2 (10 g/l, H2O, 20 °C)
  • Synonyms:
    Selenious anhydride, Selenium(IV) oxide
  • Hill Formula:
    O2Se
  • Chemical Formula:
    SeO2

 

Frequently Bought Together