Order Entry
Italy
Orders LinkContactUsLinkComponent
Selenium dioxide ≥99.999% (metals basis), Puratronic®

Selenium dioxide ≥99.999% (metals basis), Puratronic®

Toxic
Health
Environment
Selenium dioxide ≥99.999% (metals basis), Puratronic®
Certificates
Selenium dioxide ≥99.999% (metals basis), Puratronic®

2 Options Available Below

Product Details & Documents

Purity: 99.999% CAS No.: 7446-08-4 Molecular Formula: SeO2 Formula Weight: 110.96 Physical Form: Powder/Lump Odour: Odourless Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:14,8434
  • Melting Pt:340…350 °C (subl.)
  • Density:3.95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.