Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Selenium dioxide 99.999% (trace metals basis)

Toxic
Health
Environment
Selenium dioxide 99.999% (trace metals basis)
Certificates
Selenium dioxide 99.999% (trace metals basis)

2 Options Available Below

Product Details & Documents
  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:13,08508
  • Melting Pt:315 °C
  • Density:3,95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Off-white Crystalline powder
  • Total trace metal impurities:
    ≤0.001 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 145,00 €
    145,00 €
    1 * 10 g
       145,00 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 491,00 €
    491,00 €
    1 * 50 g
       491,00 €