Order Entry
Italy
ContactUsLinkComponent
Antimony(V) chloride for synthesis, Sigma-Aldrich®
Corrosive
Environment
Antimony(V) chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Environment
Antimony(V) chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (rigorous decomposition)
  • Melting Point:
    3 - 4 °C
  • Molar Mass:
    299.01 g/mol
  • Boiling Point:
    140 °C
  • Vapor Pressure:
    1.1 hPa (20 °C)
  • Density:
    2.35 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together