Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
Antimony(V) chloride, anhydrous 99%
 
undefined
 
 
 
Antimony(V) chloride, anhydrous 99%

 

  • Appearance:
    Colorless to yellow Liquid (May develop some turbidity or precipitate)
  • Assay:
    ≥98.5 %

 

 

Frequently Bought Together