Order Entry
Italy
ContactUsLinkComponent
Phosphorus(III) bromide for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Phosphorus(III) bromide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Phosphorus(III) bromide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (rigorous decomposition)
  • Melting Point:
    -40 °C
  • Molar Mass:
    270.69 g/mol
  • Boiling Point:
    173 - 175 °C (1013 hPa)
  • Vapor Pressure:
    2.9 hPa (20 °C)
  • Refractive Index:
    1.6945 (20 °C, 589 nm)
  • Density:
    2.85 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together