Order Entry
Italy
ContactUsLinkComponent
Carbon disulphide ≥99.9% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS for IR spectrophotometry, Riedel-de Haën™
Carbon disulphide ≥99.9% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS for IR spectrophotometry, Riedel-de Haën™
Catalog # 31627-1L
CAS Number:  
undefined
Carbon disulphide ≥99.9% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS for IR spectrophotometry, Riedel-de Haën™
Catalog # 31627-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Vapor Density:
    2.67 (vs air)
  • Explosion Limit:
    50 %
  • Assay (GC):
    Min. 99.9 %
  • Boiling range:
    46 - 47 °C
  • Density (D 20/4):
    1.262 - 1.263
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • H₂S + foreign org. S-comp.:
    Max. 0.0001 %
  • Sulfate (SO₄):
    Max. 0.00025 %
  • Sulfite (as SO₂):
    Max. 0.001 %
  • Benzene:
    Max. 0.001 %
  • Iodine consuming matters:
    complying
  • APHA:
    Max. 10
  • Autoignition Temperature:
    212 °F

Specification Test Results

About this item

 :
 :76.14 g/mol
Merck Index:13,01821
Boiling Pt:46 °C (1.013 hPa)
Melting Pt:-76 °C
Melting Pt:–76 °C
Density:1.264 g/cm³ (20 °C)
Flash Pt:-30 °C
Flash Pt:–30 °C
EINECS:200-843-6
UN:1131
ADR:3,I
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321