
Silica gel, Pore size: 60 Å ≥99.5% (metals basis) 0.040-0.063 mm (230-400 mesh)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Merck Index:14,8493
EINECS:231-545-4
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together