Order Entry
Italy
ContactUsLinkComponent
Orthoboric acid 99.5% for molecular biology DNase-, RNase-, protease-free
Health
Orthoboric acid 99.5% for molecular biology DNase-, RNase-, protease-free
Certificates
undefined
Health
Orthoboric acid 99.5% for molecular biology DNase-, RNase-, protease-free

Specification Test Results

  • Appearance:
    White Powder
  • Assay:
    ≥99.4 %
  • Dnase,Rnase,Protease activity:
    none detected
  • Chloride (Cl):
    ≤10 ppm
  • Phosphate (PO4):
    ≤10 ppm
  • Sulfate (SO4):
    ≤100 ppm
  • Arsenic (As):
    ≤1 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤50 ppm

Specification Test Results

Related Information

Frequently Bought Together