Order Entry
Italy
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Nicotine solution

Supelco® Organic Reference Standard, (−)-Nicotine solution

(−)-Nicotine solution, 1,0 mg/ml, Analytical standard, for drug analysis, 1 mL
Catalog # N5511-1ML
Supplier:   Merck
Supelco® Organic Reference Standard, (−)-Nicotine solution
Catalog # N5511-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, (−)-Nicotine solution
Supelco® Organic Reference Standard, (−)-Nicotine solution

(−)-Nicotine solution, 1,0 mg/ml, Analytical standard, for drug analysis, 1 mL

CAS Number:  
Price
190,00 €
Log in for availability
1 * 1 mL
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • GHS signal: GHS06, GHS08

Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311

Documents

Specifications

Specifications

  • Concentration:
    1,0 mg/ml
  • Pk:
    1 mL
  • Standards:
    Analytical standard, for drug analysis
  • Description:
    (−)-Nicotine solution
  • Cat. No.:
    N5511-1ML
  • M-Clarity quality level:
    MQ200
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • MDL No.:
    MFCD00006369
  • Flash point:
    9,7 °C
  • Molecular weight:
    162,23
  • CAS No.:
    54-11-5
  • Shipping conditions:
    STD

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.