Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl nitrite
Ethyl nitrite 10% in xylene
Ethyl nitrite 10% in xylene
Ethyl nitrite 10% in xylene
8.18982.9045
:
Merck
:
Ethyl nitrite 10% in xylene
8.18982.9045
:
Merck
:
Pk:
45 kg
Seq:
4388
:
:
75.07 g/mol
:
203-722-6
:
1194
:
3,I
:
109-95-5
:
C₂H₅NO₂