
Dibutyl phosphonate P 14.5-16%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1809-19-4
:
Formula:C₈H₁₉O₃P
MDL Number:MFCD00066633
:194.21 g/mol
Boiling Pt:118…119 °C (11 mmHg)
Melting Pt:–57 °C
Density:0.995 g/cm³
Flash Pt:121 °C
EINECS:217-316-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4225 to 1.4255 (20°C, 589 nm)
- Assay:14.5 to 16 % (P)
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together