

1,4-Butanediol diglycidyl ether 60%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2425-79-8
:
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00005146
:202.25 g/mol
Boiling Pt:266 °C (760 mmHg)
Density:1.1 g/cm³
Flash Pt:110 °C
EINECS:219-371-7
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4590 to 1.4650 (20°C, 589 nm)
- GC:≥55 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together