Barium nitrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10022-31-8
:
Formula:Ba(NO₃)₂
MDL Number:MFCD00003442
Merck Index:14,983
Melting Pt:592 °C (dec.)
Density:3.24
EINECS:233-020-5
UN:1446
ADR:5.1,II
:261.34 g/mol
:GHS03
:GHS07
Storage Temperature:Ambient
Frequently Bought Together