
Dilithium zirconium trioxide ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12031-83-3
:
Formula:Li₂O₃Zr
MDL Number:MFCD00016182
:153.1 g/mol
EINECS:234-760-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together