
Sodium hydroxide, pellets Ph. Eur., J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Appearance of solution:passes test
- Assay:97.0 - 100.5 %
- Carbonates:max. 2.0 %
- Chlorides:max. 200 ppm
- Heavy Metals:max. 20 ppm
- Identification:passes test
- Iron:max. 10 ppm
- Sulphates:max. 200 ppm
- NF Insoluble substances and organic matters:passes test
- NF Potassium:passes test
Specification Test Results
Frequently Bought Together