

Potassium superoxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12030-88-5
:
Formula:KO₂
MDL Number:MFCD00082411
:71.1 g/mol
Melting Pt:400 °C
Density:2,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:234-746-5
UN:2466
ADR:5.1,I
:GHS05
:GHS03
Specification Test Results
- Appearance:Yellow Powder or chunks
- Oxygen (O):≥215 cm³/g O2-content
Specification Test Results
Frequently Bought Together