Order Entry
Italy
ContactUsLinkComponent
Sodium azide for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
Sodium azide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
Environment
Sodium azide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    420 g/l (17 °C)
  • Melting Point:
    275 °C (decomposition)
  • Molar Mass:
    65.01 g/mol
  • Boiling Point:
    300 °C (rigorous decomposition)
  • Density:
    1.85 g/cm3 (20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together