Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium fluoride

Lithium fluoride

Plastic bottle for solids, 100 g
Catalog # 1.05690.0100
Irritant
Supplier:   Merck
Lithium fluoride
Catalog # 1.05690.0100
Supplier:  Merck
CAS Number:  7789-24-4
Lithium fluoride

Plastic bottle for solids, 100 g

CAS Number:  7789-24-4
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7789-24-4
  • Formula:FLi
  • MDL Number:MFCD00011090
  • Molecular Weight:25.94 g/mol
  • Merck Index:13,05553
  • Boiling Pt:1676 °C (1013 hPa)
  • Melting Pt:870 °C
  • Density:2.64 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-152-0
  • UN:3288
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.