Lithium fluoride 97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 7789-24-4 Molecular Formula: LiF Formula Weight: 25.94 Refractive Index: 1.3915 Odour: Odourless Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090
- CAS Number:7789-24-4
- Formula:FLi
- MDL Number:MFCD00011090
- Molecular Weight:25.94 g/mol
- Merck Index:14,5531
- Boiling Pt:1680 °C
- Melting Pt:845 °C
- Density:2.635
- Storage Temperature:Ambient
- EINECS:232-152-0
- UN:3288
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 7789-24-4 Molecular Formula: LiF Formula Weight: 25.94 Refractive Index: 1.3915 Odour: Odourless Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090
- CAS Number:7789-24-4
- Formula:FLi
- MDL Number:MFCD00011090
- Molecular Weight:25.94 g/mol
- Merck Index:14,5531
- Boiling Pt:1680 °C
- Melting Pt:845 °C
- Density:2.635
- Storage Temperature:Ambient
- EINECS:232-152-0
- UN:3288
- ADR:6.1,III



