

Lithium perchlorate, anhydrous ≥95.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-03-9
:
Formula:ClLiO₄
MDL Number:MFCD00011079
:106.39 g/mol
Merck Index:14,5539
Boiling Pt:400 °C (dec.)
Melting Pt:236 °C
Density:2.43
Storage Temperature:Ambient
EINECS:232-237-2
UN:1481
ADR:5.1,II
:GHS03
:GHS07
Related Information
Frequently Bought Together