Beakers, low form, BOLA
Supplier: Bohlender
Jump to:
About this item
PFA, translucent, with spout.
- Space-saving stackable design
- High thermal resistance from -200 to +250 °C
- Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.
Certifications: FDA conform.
6 Options Available Below
Product Details & Documents
PFA, translucent, with spout.
- Space-saving stackable design
- High thermal resistance from -200 to +250 °C
- Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.
Certifications: FDA conform.
Product Family Options
Product Information
- CapacityHeightExt.ØAvailabilityPrice