
Lithium chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Merck Index:14,5528
Boiling Pt:1325…1360 °C
Melting Pt:605 °C
Density:2.068
:ampouled under argon
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together