Order Entry
Italy
ContactUsLinkComponent
Potassium chloride, Supelco®
Potassium chloride, Supelco®
Catalog # 60136-250ML
Supplier:  Merck
CAS Number:  
undefined
Potassium chloride, Supelco®
Catalog # 60136-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360