Order Entry
Italy
Orders LinkContactUsLinkComponent
Potassium chloride, Supelco®

Potassium chloride, Supelco®

Glass bottle, 250 mL
Catalog # 60134-250ML-F
Supplier:   Merck
Potassium chloride, Supelco®
Catalog # 60134-250ML-F
Supplier:  Merck
CAS Number:  7447-40-7
Potassium chloride, Supelco®
Potassium chloride, Supelco®

Glass bottle, 250 mL

CAS Number:  7447-40-7
Price
127,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  Potassiumchloride
  • CAS Number:7447-40-7
  • Formula:KCl
  • MDL Number:MFCD00011360
  • Molecular Weight:74.55 g/mol
  • Merck Index:14,07621
  • Boiling Pt:1420 °C (1013 hPa)
  • Melting Pt:773 °C
  • Density:1.98 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:231-211-8
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.