Order Entry
Italy
Orders LinkContactUsLinkComponent
Sodium carbonate ≥99%, GPR RECTAPUR®

Sodium carbonate ≥99%, GPR RECTAPUR®

Supplier:   VWR Chemicals
Sodium carbonate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Sodium carbonate ≥99%, GPR RECTAPUR®

3 Options Available Below

Product Details & Documents
Synonyms:  Calcined soda, Carbonic acid disodium salt
  • CAS Number:497-19-8
  • Formula:Na₂CO₃
  • MDL Number:MFCD00003494
  • Molecular Weight:105.99 g/mol
  • Merck Index:14,08596
  • Boiling Pt:1600 °C (1013 hPa)
  • Melting Pt:854 °C
  • Density:2.53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-838-8
  • REACH:01-2119485498-19
Specifications for Product Family

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 20 ppm
Product Family Options

Product Information