Sodium carbonate, anhydrous, EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Calcined soda, Carbonic acid disodium salt
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
Boiling Pt:1600
Melting Pt:854
Density:2.53 g/cm³ (20 °C)
Molecular Weight:105.99 g/mol
Merck Index:14,08596
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:220 g/l (20 °C)
- Melting Point:854 °C
- Molar Mass:105.99 g/mol
- Bulk Density:1100 kg/m3
- Boiling Point:1600 °C (decomposition)
- Density:2.53 g/cm3 (20 °C)
- pH:11.5 (50 g/l, H2O, 25 °C)
Specification Test Results
Related Information
Frequently Bought Together