Lithium carbonate ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 554-13-2 Molecular Formula: Li2CO3 Formula Weight: 73.90 Physical Form: Powder Density: 2.11 Appearance: White Odour: Odourless pH: 10-11.5 Melting Point: 723° Boiling Point: 1310°dec. Solubility: Soluble in water and dilute acids. Insoluble in alcohol Merck Reference: 14,5527 MDL No.: MFCD00011084
- CAS Number:554-13-2
- Formula:Li₂CO₃
- MDL Number:MFCD00011084
- Molecular Weight:73.89 g/mol
- Merck Index:14,5527
- Boiling Pt:1310 °C (dec.)
- Melting Pt:723 °C
- Density:2.11
- Storage Temperature:Ambient
- EINECS:209-062-5
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 554-13-2 Molecular Formula: Li2CO3 Formula Weight: 73.90 Physical Form: Powder Density: 2.11 Appearance: White Odour: Odourless pH: 10-11.5 Melting Point: 723° Boiling Point: 1310°dec. Solubility: Soluble in water and dilute acids. Insoluble in alcohol Merck Reference: 14,5527 MDL No.: MFCD00011084
- CAS Number:554-13-2
- Formula:Li₂CO₃
- MDL Number:MFCD00011084
- Molecular Weight:73.89 g/mol
- Merck Index:14,5527
- Boiling Pt:1310 °C (dec.)
- Melting Pt:723 °C
- Density:2.11
- Storage Temperature:Ambient
- EINECS:209-062-5



