Order Entry
Italy
ContactUsLinkComponent
 
Sodium bromide ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 27739.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium bromide ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 27739.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    Min. 99 %
  • Acidity:
    Max. 0.002 meq/g
  • Alkalinity:
    Max. 0.004 meq/g
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (150°C):
    Max. 2 %
  • BrO3 (Bromate):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 0.3 %
  • NH4 (Ammonium):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Ca (Calcium):
    Max. 0.02 %
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 0.1 %
  • Mg (Magnesium):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :102.89 g/mol
Merck Index:13,08666
Boiling Pt:1396 °C (1013 hPa)
Melting Pt:753 °C
Density:3.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-599-9
REACH:01-2119490106-41
CAS Number:7647-15-6
Formula:NaBr
MDL Number:MFCD00003475