

Lithium tetrafluoroborate for synthesis, Sigma-Aldrich®
Catalog # 8.14347.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:324 °C (decomposition)
- Molar Mass:93.74 g/mol
Specification Test Results
About this item
:
:93.74 g/mol
Density:0.852 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:238-178-9
:GHS05
:GHS07
CAS Number:14283-07-9
Formula:BF₄Li
MDL Number:MFCD00011087