

Lithium tetrafluoroborate, anhydrous 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14283-07-9
:
Formula:BF₄Li
MDL Number:MFCD00011087
:93.74 g/mol
Melting Pt:275 °C (dec.)
Density:0.852 g/cm³
Storage Temperature:Ambient
EINECS:238-178-9
UN:3260
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:White to grey Crystalline powder or crystals
- Elemental Analysis:≥97.5 %
Specification Test Results
Frequently Bought Together