Order Entry
Italy
ContactUsLinkComponent
di-Sodium tetraborate hydrate (∼ 85% Na₂B₄O₇) for analysis, Supelco®
Health
Irritant
di-Sodium tetraborate hydrate (∼ 85% Na₂B₄O₇) for analysis, Supelco®
Supplier:  Merck
Certificates
Health
Irritant
di-Sodium tetraborate hydrate (∼ 85% Na₂B₄O₇) for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Assay (acidimetric):
    ≥ 85.0 %
  • Chloride (Cl):
    ≤ 0.002 %
  • Phosphate (PO₄):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Heavy metals (as Pb):
    ≤ 0.002 %
  • Ca (Calcium):
    ≤ 0.01 %
  • Fe (Iron):
    ≤ 0.001 %

Specification Test Results

Related Information

Frequently Bought Together