Order Entry
Italy
ContactUsLinkComponent
Methylcyclopentane
Methylcyclopentane
Catalog # EHERCA15085040
Supplier:  EHRENSTORFER
CAS Number:  
Methylcyclopentane
Catalog # EHERCA15085040
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :84.16 g/mol
Boiling Pt:71.8 °C (1013 hPa)
Boiling Pt:71,8 °C (1013 hPa)
Melting Pt:–142 °C
Density:0,74 g/cm³ (20 °C)
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:202-503-2
UN:2298
ADR:3,II
CAS Number:96-37-7
Formula:C₆H₁₂
MDL Number:MFCD00001382