Skip to Header
Skip to Main Content
Skip to Footer
Ricerca avanzata
Ricerca avanzata
Scheda di sicurezza
Certificati
Dati del produttore
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-tert-Butylcyclohexanone
4-tert-Butylcyclohexanone ≥99%
4-tert-Butylcyclohexanone ≥99%
4-tert-Butylcyclohexanone ≥99%
A15612.22
:
Thermo Fisher Scientific
:
4-tert-Butylcyclohexanone ≥99%
A15612.22
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
154.25 g/mol
:
113…116 °C (20 mmHg)
:
47…51 °C
:
0.893
:
96 °C (204 °F)
:
Ambient
:
202-678-5
:
98-53-3
:
C₁₀H₁₈O
:
MFCD00001642