

4-tert-Butylcyclohexanone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-53-3
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00001642
:154.25 g/mol
Boiling Pt:113…116 °C (20 mmHg)
Melting Pt:48…51 °C
Density:0,893 g/cm³ (50 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:202-678-5
:GHS07
:GHS09
Specification Test Results
- Appearance:White to light yellow Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:46 to 51 °C
- GC:≥98.5 %
- Water:≤1.0 % (K.F.)
- Anticaking agent:0.5 % (SiO2)
Specification Test Results
Frequently Bought Together