Order Entry
Italy
ContactUsLinkComponent
2-Chloropropyldimethylammonium chloride, Sigma-Aldrich®
Irritant
2-Chloropropyldimethylammonium chloride, Sigma-Aldrich®
Catalog # 8.03240.0250
Supplier:  Merck
CAS Number:  
Irritant
2-Chloropropyldimethylammonium chloride, Sigma-Aldrich®
Catalog # 8.03240.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    2000 g/l (20 °C)
  • Melting Point:
    188 - 191 °C
  • Molar Mass:
    158.07 g/mol
  • Bulk Density:
    350 kg/m3
  • Flash Point:
    220 °C
  • pH:
    5 - 6 (500 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :158.07 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:224-971-7
 :GHS07
Storage Temperature:Ambient
CAS Number:4584-49-0
Formula:C₅H₁₃Cl₂N
MDL Number:MFCD00012534