Order Entry
Italy
ContactUsLinkComponent
2-Chloroethyldimethylammonium chloride, Sigma-Aldrich®
Tossico
2-Chloroethyldimethylammonium chloride, Sigma-Aldrich®
Nome del fornitore:  Merck
Certificati
Tossico
2-Chloroethyldimethylammonium chloride, Sigma-Aldrich®
Nome del fornitore:  Merck

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Risultati dei test sulle specifiche

  • Solubility:
    2000 g/l (20 °C)
  • Melting Point:
    205 - 208 °C
  • Molar Mass:
    144.04 g/mol
  • Ignition Point:
    220 °C

Risultati dei test sulle specifiche

Informazioni correlate

Frequently Bought Together