Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dipropyl phthalate
Dipropyl phthalate
Dipropyl phthalate
Dipropyl phthalate
EHERC16177000
:
EHRENSTORFER
:
Dipropyl phthalate
EHERC16177000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
250.29 g/mol
:
205-015-8
:
3082
:
9,III
:
131-16-8
:
C₁₄H₁₈O₄
:
MFCD00009371