

2-Chloro-3-formylpyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36404-88-3
:
Formula:C₆H₄ClNO
MDL Number:MFCD01315308
:141.56 g/mol
Melting Pt:50…54 °C
Flash Pt:>110 °C (230 °F)
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together