Methyl-3-bromo-5-fluorobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:334792-52-8
:
Formula:C₈H₆BrFO₂
MDL Number:MFCD07780735
:233.04 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together